missing translation for 'onlineSavingsMsg'
Learn More

17-O-Acetyl-6-methylprednisolone, TRC

Product Code. 30011440
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011440 250 mg 250mg
30135202 50 mg 50mg
30009670 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011440 Supplier Toronto Research Chemicals Supplier No. TRCA186770250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86401-94-7
Molecular Formula C24 H32 O6
Molecular Weight (g/mol) 416.51
Synonym (6S,8S,9S,10R,11S,13S,14S,17R)-11-Hydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl Acetate
IUPAC Name [(6S,8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(=O)C)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14

Specifications

Analyte or Component Names 17-O-Acetyl-6-methylprednisolone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C24H32O6/c1-13-9-16-17-6-8-24(20(29)12-25,30-14(2)26)23(17,4)11-19(28)21(16)22(3)7-5-15(27)10-18(13)22/h5,7,10,13,16-17,19,21,25,28H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,21+,22-,23-,24-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 250 mg
Formula Weight 416.2199
Chemical Name or Material 17-O-Acetyl-6-methylprednisolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.