missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17beta-Dihydroequilin-2,4,16,16-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 17 beta-Dihydroequilin-2,4,16,16-d4 |
| CAS | 350819-99-7 |
| Molecular Formula | C18 2H4 H18 O2 |
| Quantity | 0.01 g |
| Synonym | Estra-1,3,5(10),7-tetraene-2,4,16,16-d4-3,17-diol, (17β)- (9 CI), (17β)-Estra-1,3,5(10),7-tetraene-3,17-diol-2,4,16,16-d4, 17β-Dihydroequilin-d4, 17β-Dihydroequilin-2,4,16,16-D4, 7-Dehydroestradiol-2,4,16,16-d4, β-Dihydroequilin-d4 |
| InChI Formula | InChI=1 S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16-17,19-20 H,2,6-9H2,1H3/t14-,16+,17+,18+/m1/s1/i3D,7D2,10 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@H]4 O)C3=CCc2c([2 H])c1O |
| IUPAC Name | (9 S,13 S,14 S,17 S)-2,4,16,16-tetradeuterio-13-methyl-9,11,12,14,15,17-hexahydro-6 H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 274.39 |
| Formula Weight | 274.1871 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction