missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17beta-Estradiol-2,4,16,16-d4 17-Pentanoate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Estradiol Valerate-2,4,16,16-D4 |
| CAS | 2673269-82-2 |
| Molecular Formula | C23 D4 H28 O3 |
| Quantity | 0.005 g |
| Synonym | Estradiol-2,4,16,16-d4 17-Valerate, Estradiol Valerate-2,4,16,16-D4, (17β)-Estra-1,3,5(10)-triene-3,17-diol-2,4,16,16-d4 17-pentanoate, 17β-Estradiol-2,4,16,16-d4 17-Pentanoate |
| InChI Formula | InChI=1 S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24 H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1/i7D,11D2,14 D |
| SMILES | [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@H]4 OC(=O)CCCC)[C@@H]3 CCc2c([2 H])c1O |
| IUPAC Name | [(8 R,9 S,13 S,14 S,17 S)-2,4,16,16-tetradeuterio-3-hydroxy-13-methyl-7,8,9,11,12,14,15,17-octahydro-6 H-cyclopenta[a]phenanthren-17-yl] pentanoate |
| Molecular Weight (g/mol) | 360.523 |
| Formula Weight | 360.2603 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction