missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-5-fluorobenzene-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,3-Dibromo-5-fluorobenzene-D3 |
| CAS | 1219805-87-4 |
| Molecular Formula | C6 D3 Br2 F |
| Quantity | 1 g |
| Synonym | 1,3-Dibromo-5-fluorobenzene-d3, 2,4-Dibromo-6-fluorobenzene-1,3,5-d3 |
| InChI Formula | InChI=1 S/C6H3Br2F/c7-4-1-5(8)3-6(9)2-4/h1-3 H/i1D,2 D,3 D |
| SMILES | [2 H]c1c(F)c([2 H])c(Br)c([2 H])c1Br |
| IUPAC Name | 1,3-dibromo-2,4,6-trideuterio-5-fluorobenzene |
| Molecular Weight (g/mol) | 256.9129 |
| Formula Weight | 254.8774 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction