Learn More
4-Bromoanisole-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromoanisole-2,3,5,6-D4 |
| CAS | 152404-45-0 |
| Molecular Formula | C7 D4 H3 Br O |
| Quantity | 1 g |
| Synonym | Benzene-1,2,4,5-d4, 3-bromo-6-methoxy- (9 CI), 1-Bromo-4-methoxy-2,3,5,6-tetradeuteriobenzene, 4-Bromoanisole-d4, 4-Bromoanisole-2,3,5,6-d4, 4-Bromoanisole-2,3,5,6-D4, 1-Bromo-4-methoxybenzene-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H7BrO/c1-9-7-4-2-6(8)3-5-7/h2-5 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(OC)c([2 H])c([2 H])c1Br |
| IUPAC Name | 1-bromo-2,3,5,6-tetradeuterio-4-methoxybenzene |
| Molecular Weight (g/mol) | 191.0585 |
| Formula Weight | 189.9931 g/mol |
| Show More |