Learn More
Benzonitrile-d5, CDN
Product Code. 30325271
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325271
Brand: CDN CDND02921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzonitrile-D5
CAS 2102-15-0
Molecular Formula C7 D5 N
Quantity 1 g
Synonym Benzonitrile-2,3,4,5,6-d5 (ACI), Benzonitrile-d5 (7 CI, 8 CI, 9 CI), Cyanobenzene-d5, Benzonitrile-D5
InChI Formula InChI=1 S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5 H/i1D,2 D,3 D,4 D,5 D
SMILES [2 H]c1c([2 H])c([2 H])c(C#N)c([2 H])c1[2 H]
IUPAC Name 2,3,4,5,6-pentadeuteriobenzonitrile
Molecular Weight (g/mol) 108.152
Formula Weight 108.074 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity