missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl-d6-amine HCl, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Dimethylamine-d6 Hydrochloride |
| CAS | 53170-19-7 |
| Molecular Formula | C2 2H6 H N . Cl H |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])NC([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1-trideuterio-N-(trideuteriomethyl)methanamine;hydrochloride |
| Molecular Weight (g/mol) | 87.58 |
| Formula Weight | 87.0722 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction