missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate-d3, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Ethyl acetate 2,2,2-d3 |
| CAS | 90691-33-1 |
| Molecular Formula | C4 2H3 H5 O2 |
| Quantity | 1 g |
| Synonym | Acetic-2,2,2-d3 acid, ethyl ester, Acetic-d3 acid, ethyl ester (9 CI), Ethyl acetate-d3 (6 CI), Acetic-2,2,2-d3 acid ethyl ester, Ethyl acetate-2,2,2-D3 |
| InChI Formula | InChI=1 S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)OCC |
| IUPAC Name | ethyl 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 91.12 |
| Formula Weight | 91.0713 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction