missing translation for 'onlineSavingsMsg'
Learn More

Vildagliptin Amide, TRC

Product Code. 17981195
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981195 1 g 1g
17943955 50 mg 50mg
17990445 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981195 Supplier Toronto Research Chemicals Supplier No. TRCH9713801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565453-39-6
Synonym N-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)glycyl-L-prolinamide∣Vildagliptin Amide∣Vildagliptin Impurity A

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 321.41
Product Type Metabolite
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.