missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl Alcohol-d10, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1-Butanol D10 |
| CAS | 34193-38-9 |
| Molecular Formula | C4 2H10 O |
| Quantity | 1 g |
| Synonym | 1-Butan-1,1,2,2,3,3,4,4,4-d9-ol-d (9 CI), Butyl-d9 alcohol-d (8 CI), 1-Butanol-d10, Butanol-d10, Decadeutero-1-butanol, Perdeutero-1-butanol, 1-Butanol D10 |
| InChI Formula | InChI=1 S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i1D3,2D2,3D2,4D2,5 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4-nonadeuterio-4-deuteriooxybutane |
| Molecular Weight (g/mol) | 84.18 |
| Formula Weight | 84.1359 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction