Learn More
Propionyl-d5 Chloride, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Propionyl-D5 Chloride |
| CAS | 352439-04-4 |
| Molecular Formula | C3 2H5 Cl O |
| Quantity | 1 g |
| Synonym | Propanoyl-d5 chloride (9 CI), Propionyl-d5 chloride, Propionyl-D5 chloride, Propionic acid chloride-d5, Propionyl chloride-d5, Propionyl-2,2,3,3,3-d5 chloride |
| InChI Formula | InChI=1 S/C3H5ClO/c1-2-3(4)5/h2H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)Cl |
| IUPAC Name | 2,2,3,3,3-pentadeuteriopropanoyl chloride |
| Molecular Weight (g/mol) | 97.555 |
| Formula Weight | 97.034 g/mol |
| Show More |