Learn More
Trimethyl-d9-amine HCl, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Trimethylamine-d9 Hydrochloride |
| CAS | 18856-86-5 |
| Molecular Formula | C3 2H9 N . Cl H |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1 H/i1D3,2D3,3D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1-trideuterio-N,N-bis(trideuteriomethyl)methanamine;hydrochloride |
| Molecular Weight (g/mol) | 104.63 |
| Formula Weight | 104.1067 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |