missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-nonanol, 98%, Thermo Scientific™

Product Code. 15449039
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449039 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15449039 Supplier Thermo Scientific Alfa Aesar Supplier No. L16600.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H-Perfluoro-1-nonanol is used to produce other chemicals. For example, it is used to produce Trifluoro-methanesulfonic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-nonyl ester. This reaction needs reagent Pyridine and
TRUSTED_SUSTAINABILITY

Specifications

CAS 423-56-3
Melting Point 66°C to 71°C
Boiling Point 176°C
Molecular Formula C9H3F17O
Linear Formula CF3(CF2)7CH2OH
MDL Number MFCD00153183
Quantity 5 g
Beilstein 1809099
Synonym 1h,1h-perfluoro-1-nonanol, 1h,1h-perfluorononanol, 1h,1h-heptadecafluoro-1-nonanol, 1h,1h-perfluorononan-1-ol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-1-nonanol, heptadecafluorooctylmethanol, 1h,1h-heptadecafluorononanol, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro, 1-nonanol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-1-nonanol #
Solubility Information Insoluble in water.
InChI Key BSXJTDJJVULBTQ-UHFFFAOYSA-N
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononan-1-ol
Molecular Weight (g/mol) 450.10
PubChem CID 533991
Formula Weight 450.1
Percent Purity 98%
Chemical Name or Material 1H,1H-Perfluoro-1-nonanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.