missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluorooctyl methacrylate, 97%, Thermo Scientific™

Product Code. 11358435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358435 1 g 1g
11368435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358435 Supplier Thermo Scientific Alfa Aesar Supplier No. L10440.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3934-23-4
Molecular Formula C12H7F15O2
Molecular Weight (g/mol) 468.162
MDL Number MFCD00039247
InChI Key RUEKTOVLVIXOHT-UHFFFAOYSA-N
Synonym 1h,1h-perfluorooctyl methacrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl methacrylate, 1h,1h-perfluorooctyl methacrylate, contains mehq as inhibitor, 2-propenoic acid,2-methyl-, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester
PubChem CID 77532
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.496
Boiling Point 70°C (1 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.337
Quantity 1 g
Beilstein 6445701
Formula Weight 468.16
Percent Purity 97%
Chemical Name or Material 1H,1H-Perfluorooctyl methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.