missing translation for 'onlineSavingsMsg'
Learn More

1H,4H,6H,7H-Pyrano[4,3-c]pyrazol-3-ylmethanamine, TRC

Product Code. 30399610
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399610 500 mg
30406420 100 mg
30403594 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399610 Supplier Toronto Research Chemicals Supplier No. TRCH160086500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933722-15-7
Molecular Formula C7H11N3O
Molecular Weight (g/mol) 153.18
Synonym 2,4,6,7-Tetrahydropyrano[4,3-c]pyrazole-3-methanamine
IUPAC Name 2,4,6,7-tetrahydropyrano[4,3-c]pyrazol-3-ylmethanamine
SMILES NCc1[nH]nc2CCOCc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H11N3O/c8-3-7-5-4-11-2-1-6(5)9-10-7/h1-4,8H2,(H,9,10)
Formula Weight 153.0902
Chemical Name or Material 1H,4H,6H,7H-Pyrano[4,3-c]pyrazol-3-ylmethanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.