missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazol-4-amine, 97%, Thermo Scientific™

Product Code. 10391750
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391750 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391750 Supplier Thermo Scientific Maybridge Supplier No. MAY00334DA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 41748-71-4
Assay Percent Range 97%
Molecular Formula C7H7N3
MDL Number MFCD03305710
Quantity 1 g
Synonym 4-amino-1h-indazole, 4-aminoindazole, 2h-indazol-4-amine, 1h-indazole-4-ylamine, 1h-indazol-4-ylamine, 4-amino indazole, 4-amino-indazole, 1h-indazole-4-amine, pubchem20573, indazol-4-amine
InChI Key MDELYEBAXHZXLZ-UHFFFAOYSA-N
SMILES NC1=C2C=NNC2=CC=C1
IUPAC Name 1H-indazol-4-amine
Molecular Weight (g/mol) 133.15
PubChem CID 413085
Formula Weight 133.15
Chemical Name or Material 1H-Indazol-4-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.