missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-1-carboxamidine Hydrochloride, TRC

Product Code. 30413504
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413504 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413504 Supplier Toronto Research Chemicals Supplier No. TRCP84250025G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-pyrazole-1-carboximidamide
CAS 4023-02-3
Molecular Formula C4 H6 N4 . Cl H
Quantity 25 g
Synonym 1H-Pyrazole-1-carboximidamide','hydrochloride (1:1), 1H-Pyrazole-1-carboximidamide','monohydrochloride (9CI), Pyrazole-1-carboxamidine','hydrochloride (7CI,8CI), 1-Amidinopyrazole hydrochloride, 1H-Pyrazole-1-carboxamidine hydrochloride, Praxadine hydrochloride, Pyrazole-1-carboximidamide hydrochloride, 1H-Pyrazole-1-carboximidamide hydrochloride, Zanamivir Imp. F (EP) as Hydrochloride, PCH, GR 169473A, Ph Eur Zanamivir Impurity F, GR 152919X, GR 169473A, GR 212339X, GR 228446X, USP Zanamivir Related Compounds Mixture, PCH Test Mix 1
InChI Formula InChI=1S/C4H6N4.ClH/c5-4(6)8-3-1-2-7-8;/h1-3H,(H3,5,6);1H
SMILES Cl.NC(=N)n1cccn1
IUPAC Name pyrazole-1-carboximidamide;hydrochloride
Molecular Weight (g/mol) 146.58
Formula Weight 146.0359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.