missing translation for 'onlineSavingsMsg'
Learn More

1H-Tetrazole (~0.3M in Acetonitrile), TRC

Product Code. 30449351
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449351 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449351 Supplier Toronto Research Chemicals Supplier No. TRCT307505100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Tetrazole
CAS 288-94-8
Molecular Formula C H2 N4
Quantity 100 mL
Synonym 2H-Tetrazole, 1H-TZ, 2,3,4-Triazapyrrole, NSC 36712, Tetraazacyclopentadiene
InChI Formula InChI=1S/CH2N4/c1-2-4-5-3-1/h1H,(H,2,3,4,5)
SMILES N1=NNC=N1
IUPAC Name 1H-tetrazole
Molecular Weight (g/mol) 70.06
Formula Weight 70.03
Concentration or Composition (by Analyte or Components) ~0.3 M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.