missing translation for 'onlineSavingsMsg'
Learn More

1H-Tetrazole, 0.45M in acetonitrile

Product Code. 15473699
Change view
Click to view available options
Quantity:
45 mL
200 mL
450 mL
Unit Size:
200mL
450mL
45mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473699 450 mL 450mL
15483699 45 mL 45mL
15463699 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15473699 Supplier Thermo Scientific Alfa Aesar Supplier No. J61034.AO
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Coupling reagent for preparation of polynucleotides | CAS: 288-94-8 | CH2N4 | 70.055 g/mol

1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

Solubility
Miscible with water.

Notes
Incompatible with reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-94-8
Molecular Formula CH2N4
Molecular Weight (g/mol) 70.055
MDL Number MFCD00005247
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
PubChem CID 67519
ChEBI CHEBI:33193
IUPAC Name 2H-tetrazole
SMILES C1=NNN=N1

Specifications

Density 0.798
Quantity 450 mL
UN Number UN1648
Beilstein 105799
Merck Index 14,2786
Synonym 1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
Solubility Information Miscible with water.
Formula Weight 70.05
Concentration or Composition (by Analyte or Components) 0.45M in acetonitrile
Physical Form Liquid
Chemical Name or Material 1H-Tetrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.