missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deuterated PAH Mixture 918 200 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Acenaphthene D10∣Benzo(a)pyrene D12∣Benzo[a]anthracene D12∣Dibenzo(a,h)anthracene D14∣Dibenzo[a,i]pyrene-d14∣Fluoranthene D10∣Phenanthrene D10 |
| CAS | 15067-26-2, 63466-71-7, 1718-53-2, 13250-98-1, 158776-07-9, 93951-69-0 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acenaphthene D10 200 μg/mL∣Benzo(a)pyrene D12 200 μg/mL∣Benzo[a]anthracene D12 200 μg/mL∣Dibenzo(a,h)anthracene D14 200 μg/mL∣Dibenzo[a,i]pyrene-d14 200 μg/mL∣Fluoranthene D10 200 μg/mL∣Phenanthrene D10 200 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction