Learn More
(1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 39961-95-0 |
| Molecular Formula | C10H20N2O6 |
| MDL Number | MFCD00191979 |
| Quantity | 10 g |
| SMILES | N[C@H]1[C@H](N)CCCC1.O=C(O)[C@H](O)[C@@H](O)C(O)=O |
| Molecular Weight (g/mol) | 264.28 |
| Percent Purity | 95% |
| Chemical Name or Material | (1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate |