missing translation for 'onlineSavingsMsg'
Learn More

(1R,5S,6R)-Ethyl 5-(Pentan-3-yloxy)-7-oxabicyclo[4.1.0]hept-3-ene-3-carboxylate, TRC

Product Code. 30377974
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377974 2.5 mg 2.5mg
30387301 25 mg -
30385691 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377974 Supplier Toronto Research Chemicals Supplier No. TRCE9179902.5MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125971-95-1
Molecular Formula C40 H47 F N2 O5
Molecular Weight (g/mol) 654.81
Synonym 1,1-Dimethylethyl (4R-cis)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1-H-pyrrol-1-yl]ethyl]-2,2-di-methyl-1,3-dioxane-4-acetate
IUPAC Name tert-butyl 2-[(4R,6R)-6-[2-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O5

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C40H47FN2O5/c1-26(2)36-35(38(45)42-30-16-12-9-13-17-30)34(27-14-10-8-11-15-27)37(28-18-20-29(41)21-19-28)43(36)23-22-31-24-32(47-40(6,7)46-31)25-33(44)48-39(3,4)5/h8-21,26,31-32H,22-25H2,1-7H3,(H,42,45)/t31-,32-/m1/s1
Formula Weight 654.3469
Chemical Name or Material 1,1-Dimethylethyl (4R-cis)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1-H-pyrrol-1-yl]ethyl]-2,2-di-methyl-1,3-dioxane-4-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.