Learn More
(1R)-(+)-a-Pinene (~80% ee), TRC
Product Code. 30135813
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135813 10 g 10g
30133280 50 g 50g
30004280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135813 Supplier Toronto Research Chemicals Supplier No. TRCP46661510G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7785-70-8
Molecular Formula C10 H16
Molecular Weight (g/mol) 136.23
Synonym Bicyclo[3.1.1]hept-2-ene, 2,6,6-trimethyl-, (1R,5R)-, 2-Pinene, (1R,5R)-(+)- (8CI), Bicyclo[3.1.1]hept-2-ene, 2,6,6-trimethyl-, (1R)-, (+)-(1R)-α-Pinene, (+)-(1R,5R)-α-Pinene, (+)-2-Pinene, (+)-α-Pinene, (1R)-(+)-α-Pinene, (1R)-α-Pinene, (1R,5R)-(+)-α-Pinene, (1R,5R)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (R)-(+)-α-Pinene, d-α-Pinene
IUPAC Name (1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES C1=C(C)C2CC(C1)C2(C)C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m1/s1
Formula Weight 136.1252
Chemical Name or Material (1R)-(+)-alpha-Pinene