missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-Fmoc-acpc, TRC

Product Code. 30408674
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408674 500 mg
30392838 100 mg
30392241 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408674 Supplier Toronto Research Chemicals Supplier No. TRCF601220500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 359586-64-4
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.4
Synonym Fmoc-(1S,2S)-2-aminocyclopentane carboxylic Acid
SMILES OC(=O)[C@H]1CCC[C@@H]1NC(=O)OCC2c3ccccc3c4ccccc24

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H21NO4/c23-20(24)17-10-5-11-19(17)22-21(25)26-12-18-15-8-3-1-6-13(15)14-7-2-4-9-16(14)18/h1-4,6-9,17-19H,5,10-12H2,(H,22,25)(H,23,24)/t17-,19-/m0/s1
Formula Weight 351.1471
Chemical Name or Material (1S,2S)-Fmoc-acpc
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.