missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-N-Desmethyl Sertraline Hydrochloride, TRC

Product Code. 17942085
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942085 1 mg 1mg
17913765 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942085 Supplier Toronto Research Chemicals Supplier No. TRCD2921011MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675126-10-0
Molecular Formula C16 H15 Cl2 N . Cl H
Molecular Weight (g/mol) 328.66
Synonym Norsertraline Hydrochloride, (1S,4S)-N-Desmethylsertraline Hydrochloride
IUPAC Name (1S,4S)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES Cl.N[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/t11-,16-;/m0./s1
Formula Weight 327.0348
Percent Purity >95%
Chemical Name or Material (1S,4S)-N-Desmethyl Sertraline Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.