Learn More
2-(1,2-Benzisoxazol-3-yl)acetic acid, 97%, Thermo Scientific™
Product Code. 10425712
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10425712 250 mg 250mg
10425902 1 g 1g
10220595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10425712 Supplier Thermo Scientific Maybridge Supplier No. KM10833CB
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Chemical Identifiers
CAS 4865-84-3
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD02180388
InChI Key BVSIAYQIMUUCRW-UHFFFAOYSA-M
Synonym 2-1,2-benzisoxazol-3-yl acetic acid, 1,2-benzisoxazole-3-acetic acid, 1,2-benzisoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl acetic acid, 2-benzo d isoxazol-3-yl acetic acid, 1,2-benzoxazol-3-ylacetic acid, 2-benzo d isoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl ethanoic acid, nvu, zlchem 388
PubChem CID 301729
SMILES [O-]C(=O)CC1=NOC2=CC=CC=C12
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 2-(1,2-benzoxazol-3-yl)acetic acid
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 2-(1, 2-Benzisoxazol-3-yl)acetic acid