missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10275454
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10275454 10 g 10g
10366564 250 mg 250mg
10431314 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275454 Supplier Thermo Scientific Maybridge Supplier No. CC32714EA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 26286-54-4
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD03274531
InChI Key HVECTIQVQPUSEX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
PubChem CID 320166
IUPAC Name 2-imidazol-1-ylaniline
SMILES C1=CC=C(C(=C1)N)N2C=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 2-(1H-Imidazol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.