missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2,2-Trimethylacetamido)benzeneboronic acid, 95%, Thermo Scientific™

Product Code. 11491650
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491650 1 g 1g
15457645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491650 Supplier Thermo Scientific Alfa Aesar Supplier No. H27004.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146140-95-6
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD01114645
InChI Key MXRAJVMTCAUABO-UHFFFAOYSA-N
Synonym 2-tert-butylcarbonylamino phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivaloylamino phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivaloylaminobenzene boronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
PubChem CID 4193502
IUPAC Name [2-(2,2-dimethylpropanoylamino)phenyl]boronic acid
SMILES CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O

Specifications

Melting Point 268°C to 271°C
Quantity 1 g
Formula Weight 221.06
Percent Purity 95%
Chemical Name or Material 2-(2,2,2-Trimethylacetamido)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.