missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-1-(4-nitrophenyl)ethanol, TRC

Product Code. 30457104
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30457104 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457104 Supplier Toronto Research Chemicals Supplier No. TRCT900328100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 241127-76-4
Molecular Formula C8H6NO3F3
Quantity 100 mg
InChI Formula InChI=1S/C8H6F3NO3/c9-8(10,11)7(13)5-1-3-6(4-2-5)12(14)15/h1-4,7,13H
SMILES OC(c1ccc(cc1)[N+](=O)[O-])C(F)(F)F
IUPAC Name 2,2,2-trifluoro-1-(4-nitrophenyl)ethanol
Molecular Weight (g/mol) 221.13
Formula Weight 221.03
Chemical Name or Material 2,2,2-Trifluoro-1-(4-nitrophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.