missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetaldehyde (72% in aqueous solution) Monohydrate, TRC

Product Code. 30450240
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450240 1 g -
30454292 50 g 50g
30454214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450240 Supplier Toronto Research Chemicals Supplier No. TRCT7800001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33953-86-5
Molecular Formula C2 H F3 O . H2 O
Molecular Weight (g/mol) 116.04
Synonym Fluoral Monohydrate, 2,2,2-Trifluoroacetaldehyde Monohydrate, Fluoral (aldehyde) Monohydrate, NSC 9446 Monohydrate, Trifluoroacetaldehyde Monohydrate, Trifluoroethanal Monohydrate, Trifluoromethyl Aldehyde Mnohydrate
IUPAC Name 2,2,2-trifluoroacetaldehyde;hydrate
SMILES O.FC(F)(F)C=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2HF3O.H2O/c3-2(4,5)1-6;/h1H;1H2
Formula Weight 116.0085
Concentration or Composition (by Analyte or Components) 72%
Chemical Name or Material 2,2,2-Trifluoroacetaldehyde Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.