missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Triflate, TRC

Product Code. 30452819
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30452819

Brand: Toronto Research Chemicals TRCT7903001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,2-Trifluoroethyl Triflate
CAS 6226-25-1
Molecular Formula C3H2F6O3S
Quantity 1 g
Synonym Trifluoromethanesulfonic Acid 2,2,2-Trifluoroethyl Ester, 2,2,2-Trifluoroethanol Trifluoromethanesulfonate, 2,2,2-Trifluoroethyl trifluoromethanesulfonate, 2,2,2-Trifluoroethyl trifluoromethylsulfonate
InChI Formula InChI=1S/C3H2F6O3S/c4-2(5,6)1-12-13(10,11)3(7,8)9/h1H2
SMILES FC(F)(F)COS(=O)(=O)C(F)(F)F
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
Molecular Weight (g/mol) 232.1
Formula Weight 231.9629
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.