Learn More
2,2,3,3,3-Pentafluoropropyl acrylate, 97%
Product Code. 11348385
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348385 1 g 1g
11358385 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11348385 Supplier Thermo Scientific Alfa Aesar Supplier No. L10343.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
CAS: 356-86-5 | C6H5F5O2 | 204.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 356-86-5
Molecular Formula C6H5F5O2
Molecular Weight (g/mol) 204.10
MDL Number MFCD00039257
InChI Key JDVGNKIUXZQTFD-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoropropyl acrylate, 1h,1h-pentafluoropropyl acrylate, 2-propenoic acid, 2,2,3,3,3-pentafluoropropyl ester, acmc-20aokp, acrylic acid=2,2,3,3,3-pentafluoropropyl ester, acrylic acid 2,2,3,3,3-pentafluoropropyl ester, 2-propenoic acid,2,2,3,3,3-pentafluoropropyl ester, 2,2,3,3,3-pentafluoropropyl acrylate stabilized with tbc, 2,2,3,3,3-pentafluoropropyl acrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
PubChem CID 67744
IUPAC Name 2,2,3,3,3-pentafluoropropyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)COC(=O)C=C
Specifications
Density 1.32
Boiling Point 49°C to 50°C (100 mmHg)
Flash Point 23°C (73°F)
Odor Pungent
Linear Formula H2C=CHCO2CH2CF2CF3
Refractive Index 1.336
Quantity 1 g
UN Number UN3272
Beilstein 1779140
Formula Weight 204.1
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl acrylate