missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.

Product Code. 11495767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495767 25 g 25g
11485767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495767 Supplier Thermo Scientific Alfa Aesar Supplier No. B24340.14
  • ONLINE EXCLUSIVE PRICE
    215.00€
    193.65€ / 25g
    Save 21.35€
    9% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 45115-53-5 | C7H7F5O2 | 218.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45115-53-5
Molecular Formula C7H7F5O2
Molecular Weight (g/mol) 218.123
MDL Number MFCD00039256
InChI Key CLISWDZSTWQFNX-UHFFFAOYSA-N
Synonym 1h,1h-pentafluoropropyl methacrylate, 2,2,3,3,3-pentafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3,3-pentafluoropropyl ester, 1h,1h-pentafluoro-n-propyl methacrylate, 2,2,3,3,3-pentafluoro-n-propyl methacrylate, methacrylic acid 2,2,3,3,3-pentafluoropropyl ester, 2,2,3,3,3-pentafluoropropyl methacrylate stabilized with tbc, 2,2,3,3,3-pentafluoropropyl methacrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
PubChem CID 123516
IUPAC Name 2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(F)(F)F)(F)F

Specifications

Density 1.279
Boiling Point 54°C to 55°C (100 mmHg)
Flash Point 29°C (84°F)
Refractive Index 1.348
Quantity 25 g
UN Number UN3272
Formula Weight 218.12
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl methacrylate, stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.