missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetramethylcyclopropanecarbonyl Chloride (>90%), TRC

Product Code. 30415080
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415080 2.5 g 2.5g
30412889 1 g -
30413751 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415080 Supplier Toronto Research Chemicals Supplier No. TRCT3020252.5G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24303-61-5
Molecular Formula C8H13ClO
Molecular Weight (g/mol) 160.64
IUPAC Name 2,2,3,3-tetramethylcyclopropane-1-carbonyl chloride
SMILES CC1(C)C(C(=O)Cl)C1(C)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H13ClO/c1-7(2)5(6(9)10)8(7,3)4/h5H,1-4H3
Formula Weight 160.0655
Chemical Name or Material 2,2,3,3-Tetramethylcyclopropanecarbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.