missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,6-Hexakis(2,2,3,3,4,4-hexafluorobutoxy)-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-Triazatriphosphorine, TRC

Product Code. 17964815
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964815 10 mg 10mg
17912865 5 mg 5mg
17920535 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964815 Supplier Toronto Research Chemicals Supplier No. TRCH29397810MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186406-47-3
Molecular Formula C24H18F36N3O6P3
Molecular Weight (g/mol) 1221.28
Synonym Hexakis(1H,1H,4H-hexafluorobutyloxy)phosphazine;
SMILES FC(F)C(F)(F)C(F)(F)COP1(OCC(F)(F)C(F)(F)C(F)F)=NP(OCC(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)F)=NP(OCC(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)F)=N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H18F36N3O6P3/c25-7(26)19(49,50)13(37,38)1-64-70(65-2-14(39,40)20(51,52)8(27)28)61-71(66-3-15(41,42)21(53,54)9(29)30,67-4-16(43,44)22(55,56)10(31)32)63-72(62-70,68-5-17(45,46)23(57,58)11(33)34)69-6-18(47,48)24(59,60)12(35)36/h7-12H,1-6H2
Formula Weight 1220.9834
Percent Purity >95%
Chemical Name or Material 2,2,4,4,6,6-Hexakis(2,2,3,3,4,4-hexafluorobutoxy)-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-Triazatriphosphorine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.