missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4,6,6-Hexakis(2,2,3,3-tetrafluoropropoxy)-2Lambda5,4Lambda5,6Lambda5-1,3,5,2,4,6-triazatriphosphorine, TRC

Product Code. 30134078
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134078 2.5 g 2.5g
17931985 250 mg 250mg
17940975 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134078 Supplier Toronto Research Chemicals Supplier No. TRCH2939602.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58943-98-9
Molecular Formula C18H18F24N3O6P3
Molecular Weight (g/mol) 921.23
Synonym 2,2,4,4,6,6-Hexahydro-2,2,4,4,6,6-hexakis(2,2,3,3-tetrafluoropropoxy)-1,3,5,2,4,6-triazatriphosphorine
IUPAC Name 2,2,4,4,6,6-hexakis(2,2,3,3-tetrafluoropropoxy)-1,3,5-triaza-2λ^{5},4λ^{5},6λ^{5}-triphosphacyclohexa-1,3,5-triene
SMILES FC(F)C(F)(F)COp1(OCC(F)(F)C(F)F)np(OCC(F)(F)C(F)F)(OCC(F)(F)C(F)F)np(OCC(F)(F)C(F)F)(OCC(F)(F)C(F)F)n1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C18H18F24N3O6P3/c19-7(20)13(31,32)1-46-52(47-2-14(33,34)8(21)22)43-53(48-3-15(35,36)9(23)24,49-4-16(37,38)10(25)26)45-54(44-52,50-5-17(39,40)11(27)28)51-6-18(41,42)12(29)30/h7-12H,1-6H2
Formula Weight 921.0025
Percent Purity >95%
Chemical Name or Material 2,2,4,4,6,6-Hexakis(2,2,3,3-tetrafluoropropoxy)-2lambda5,4lambda5,6lambda5-1,3,5,2,4,6-triazatriphosphorine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.