missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,4-Tetramethyl-3-pentanone, 98%, Thermo Scientific™

Product Code. 10617780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617780 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10617780 Supplier Thermo Scientific Acros Supplier No. 434760010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 815-24-7
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8240g/mL
Boiling Point 150.0°C to 155.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H18O
MDL Number MFCD00008845
Quantity 1 g
Specific Gravity 0.824
Synonym hexamethylacetone, 2,2,4,4-tetramethyl-3-pentanone, di-tert-butyl ketone, 3-pentanone, 2,2,4,4-tetramethyl, 2,2,4,4-tetramethyl-pentan-3-one, 3-pentanone,2,2,4,4-tetramethyl, di-t-butyl ketone, acmc-20akrk, t-c4h9 2co, 2,4,4-tetramethyl-3-pentanone
InChI Key UIQGEWJEWJMQSL-UHFFFAOYSA-N
SMILES CC(C)(C)C(=O)C(C)(C)C
IUPAC Name 2,2,4,4-tetramethylpentan-3-one
Molecular Weight (g/mol) 142.24
PubChem CID 13152
Formula Weight 142.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4-Tetramethyl-3-pentanone
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,4,4-Tetramethyl-3-pentanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.