missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)-1,2-propanediol, TRC

Product Code. 30137734
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137734 1 g 1g
17984365 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137734 Supplier Toronto Research Chemicals Supplier No. TRCD4460801G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118689-07-9
Molecular Formula C11 H11 F2 N3 O2
Molecular Weight (g/mol) 255.22
Synonym (2RS)-2-(2,4-Difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propane-1,2-diol, Fluconazole Imp. F (EP)
IUPAC Name 2-(2,4-difluorophenyl)-3-(1,2,4-triazol-1-yl)propane-1,2-diol
SMILES OCC(O)(Cn1cncn1)c2ccc(F)cc2F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H11F2N3O2/c12-8-1-2-9(10(13)3-8)11(18,5-17)4-16-7-14-6-15-16/h1-3,6-7,17-18H,4-5H2
Formula Weight 255.0819
Chemical Name or Material (2RS)-2-(2,4-Difluorophenyl)-3-(1H-1,2,4-triazol-1-yl)propane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.