missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 100mL, MP Biomedicals

Product Code. 11415812 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11415812

Brand: MP Biomedicals 04821809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stabilized Liquid Substrate A safe, convenient and good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions; Supplied ready-to-use at 1.46 mmol/liter

TRUSTED_SUSTAINABILITY

Specifications

Synonym ABTS
Quantity 100 mL
Chemical Name or Material 2, 2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 100mL
compliance-icons
Product Identifier
  • 2,2'-Azino-bis-(3-ethylbenzthiazoline-6-sulfonic Acid) Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-Azino-bis-(3-ethylbenzthiazoline-6-sulfonic Acid)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.