Learn More
2,2'-Binaphthalene-6,6'-dicarboxylic Acid, Mikromol™
Description
2,2'-Binaphthalene-6,6'-dicarboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 2,2'-Binaphthalene-6,6'-dicarboxylic Acid |
| CAS | 932033-58-4 |
| Analyte or Component Names | 2,2'-Binaphthalene-6,6'-dicarboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H14 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Adapalene Imp. A (EP), Adapalene USP Related Compound E, Adapalene USP RC E, 2,2'-Binaphthalene-6,6'-dicarboxylic Acid, Adapalene Related Compound E, Adapalene Impurity A |
| InChI Formula | InChI=1S/C22H14O4/c23-21(24)19-7-5-15-9-13(1-3-17(15)11-19)14-2-4-18-12-20(22(25)26)8-6-16(18)10-14/h1-12H,(H,23,24)(H,25,26) |
| SMILES | OC(=O)c1ccc2cc(ccc2c1)c3ccc4cc(ccc4c3)C(=O)O |
| IUPAC Name | 6-(6-carboxynaphthalen-2-yl)naphthalene-2-carboxylic acid |
| Show More |