missing translation for 'onlineSavingsMsg'
Learn More

2,2’-Bs(trifluoromethyl)-10H-3,10'-biphenothiazine, TRC

Product Code. 30457291
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457291 250 mg 250mg
30452669 50 mg 50mg
30453992 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457291 Supplier Toronto Research Chemicals Supplier No. TRCT790410250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2’-Bs(trifluoromethyl)-10H-3,10'-biphenothiazine
Analyte or Component Names 2,2’-Bs(trifluoromethyl)-10H-3,10'-biphenothiazine
Shipping Condition Room Temperature
Molecular Formula C26 H14 F6 N2 S2
InChI Formula InChI=1S/C26H14F6N2S2/c27-25(28,29)14-9-10-23-20(11-14)34(18-6-2-4-8-22(18)36-23)19-13-24-17(12-15(19)26(30,31)32)33-16-5-1-3-7-21(16)35-24/h1-13,33H
Solution Type Neat
SMILES FC(F)(F)c1ccc2Sc3ccccc3N(c2c1)c4cc5Sc6ccccc6Nc5cc4C(F)(F)F
IUPAC Name 2-(trifluoromethyl)-3-[2-(trifluoromethyl)phenothiazin-10-yl]-10H-phenothiazine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 532.523
Quantity 250 mg
Formula Weight 532.05
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.