missing translation for 'onlineSavingsMsg'
Learn More

2,2'-[Butane-1,4-diylbis(piperazine-1,4-diyl)]dipyrimidine, TRC

Product Code. 30132985
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132985 250 mg 250mg
30138222 50 mg 50mg
17902365 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132985 Supplier Toronto Research Chemicals Supplier No. TRCB689970250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 257877-45-5
Molecular Formula C20 H30 N8
Molecular Weight (g/mol) 382.51
Synonym 2,2'-[Butane-1,4-diylbis(piperazine-1,4-diyl)]dipyrimidine, Buspirone Hydrochloride Imp. C (EP)
IUPAC Name 2-[4-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]piperazin-1-yl]pyrimidine
SMILES C(CCN1CCN(CC1)c2ncccn2)CN3CCN(CC3)c4ncccn4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H30N8/c1(9-25-11-15-27(16-12-25)19-21-5-3-6-22-19)2-10-26-13-17-28(18-14-26)20-23-7-4-8-24-20/h3-8H,1-2,9-18H2
Formula Weight 382.2593
Chemical Name or Material 2,2'-[Butane-1,4-diylbis(piperazine-1,4-diyl)]dipyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.