missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Butoxyethoxy)ethyl Acetate, TRC

Product Code. 30348019
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348019 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348019 Supplier Toronto Research Chemicals Supplier No. TRCB69568025ML
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylene glycol-monobutylether acetate
CAS 124-17-4
Molecular Formula C10 H20 O4
Quantity 25 mL
Synonym Ethanol, 2-(2-butoxyethoxy)-, acetate (6CI,7CI,8CI,9CI), 2-(2-Butoxyethoxy)ethanol acetate, 2-(2-Butoxyethoxy)ethyl acetate, 2-(2-n-Butoxyethoxy)ethyl acetate, ABG, Butasol V, Butoxyethoxyethyl acetate, Butyl carbitol acetate, Butyl diethylene glycol acetate, Butyl diglycol acetate, DE Acetate, Diethylene glycol butyl ether acetate, Diethylene glycol mono-n-butyl ether acetate, Diethylene glycol monobutyl acetate, Diethylene glycol monobutyl ether acetate, Diglycol monobutyl ether acetate, Hykleen 340, NSC 5175, NSC 6570
InChI Formula InChI=1S/C10H20O4/c1-3-4-5-12-6-7-13-8-9-14-10(2)11/h3-9H2,1-2H3
SMILES CCCCOCCOCCOC(=O)C
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
Molecular Weight (g/mol) 204.26
Formula Weight 204.1362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.