missing translation for 'onlineSavingsMsg'
Learn More

2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine, Mikromol™

Product Code. 17931576
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931576 25 mg 25mg
17951486 100 mg 100mg
30458362 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931576 Supplier Mikromol Supplier No. MM0029.070025
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine
CAS 129083-44-9
Analyte or Component Names 2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine
Shipping Condition Room Temperature
Molecular Formula C10 H14 N8 S4
Certifications/Compliance ISO 17025
Synonym Famotidine Imp. E (EP), Famotidine USP Related Compound E, Famotidine USP RC E, 2,2'-[Disulfanediylbis-(methylenethiazole-4,2-diyl)]diguanidine, Famotidine Related Compound E, Famotidine Impurity E
InChI Formula InChI=1S/C10H14N8S4/c11-7(12)17-9-15-5(1-19-9)3-21-22-4-6-2-20-10(16-6)18-8(13)14/h1-2H,3-4H2,(H4,11,12,15,17)(H4,13,14,16,18)
SMILES NC(=Nc1nc(CSSCc2csc(N=C(N)N)n2)cs1)N
IUPAC Name 2-[4-[[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methyldisulfanyl]methyl]-1,3-thiazol-2-yl]guanidine
Molecular Weight (g/mol) 374.53
Quantity 25 mg
Formula Weight 374.0224 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.