missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15478327
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478327 5 g 5g
15468327 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478327 Supplier Thermo Scientific Alfa Aesar Supplier No. H51851.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115311-41-6
Molecular Formula C9H6N2O2S
Molecular Weight (g/mol) 206.219
MDL Number MFCD07375352
InChI Key KJHHLEKGRGFSQH-UHFFFAOYSA-N
Synonym 2-pyridin-2-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2-pyridinyl, 2-2-pyridyl thiazole-4-carboxylic acid, 2-2-pyridinyl-4-thiazolecarboxylic acid, 2-2-pyridinyl thiazole-4-carboxylic acid, 2-pyridin-2-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-2-pyridinyl, acmc-20ml6t, 2-2-pyridinyl-4-thiazolylcarboxylic acid, 2-2-pyridyl-1,3-thiazole-4-carboxylic acid
PubChem CID 15623090
IUPAC Name 2-pyridin-2-yl-1,3-thiazole-4-carboxylic acid
SMILES C1=CC=NC(=C1)C2=NC(=CS2)C(=O)O

Specifications

Quantity 5 g
Beilstein 18407886
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-(2-Pyridyl)thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.