Learn More
2,2'-Thiodiacetic Acid, TRC
Product Code. 30454242
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454242 25 g 25g
30453936 10 g 10g
30452724 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30454242 Supplier Toronto Research Chemicals Supplier No. TRCT34456025G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-93-3
Molecular Formula C4 H6 O4 S
Molecular Weight (g/mol) 150.15
Synonym Acetic acid, 2,2'-thiobis- (9CI, ACI), 2,2'-Thiobis[acetic acid] (ACI), Acetic acid, thiobis- (3CI), Acetic acid, thiodi- (6CI, 7CI, 8CI), (Carboxymethylthio)acetic acid, 2,2'-Thiodiacetic acid, 2,2'-Thiodiethanoic acid, 2,2'-Thiodiglycolic acid, 2-(Carboxymethylsulfanyl)acetic acid, 2-[(Carboxymethyl)sulfanyl]acetic acid, Dicarboxymethyl sulfide, Dimethylsulfide-α,α'-dicarboxylic acid, Mercaptodiacetic acid, NSC 28743, NSC 40469, NSC 52326, Thiodiacetic acid, Thiodiglycolic acid, β,β'-Thiodiglycolic acid
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES OC(=O)CSCC(=O)O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C4H6O4S/c5-3(6)1-9-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
Formula Weight 149.9987
Chemical Name or Material 2,2'-Thiodiacetic acid