missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethyl-1-(3-sulfonatopropyl)-indolinium-5-sulfonic Acid, Potassium Salt (90%), TRC

Product Code. 30457051
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457051 250 mg 250mg
30449144 1 g -
30453198 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457051 Supplier Toronto Research Chemicals Supplier No. TRCT797450250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 427882-78-8
Molecular Formula C14 H18 N O6 S2 . K
Molecular Weight (g/mol) 399.52
IUPAC Name potassium;2,3,3-trimethyl-1-(3-sulfonatopropyl)indol-1-ium-5-sulfonate
SMILES [K+].CC1=[N+](CCCS(=O)(=O)[O-])c2ccc(cc2C1(C)C)S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H19NO6S2.K/c1-10-14(2,3)12-9-11(23(19,20)21)5-6-13(12)15(10)7-4-8-22(16,17)18;/h5-6,9H,4,7-8H2,1-3H3,(H-,16,17,18,19,20,21);/q;+1/p-1
Formula Weight 399.0213
Chemical Name or Material 2,3,3-Trimethyl-1-(3-sulfonatopropyl)-indolinium-5-sulfonic acid potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.