missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethylindolenine, 98%

Product Code. 15433247
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433247 250 g 250g
15423247 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433247 Supplier Thermo Scientific Alfa Aesar Supplier No. B22410.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1640-39-7 | C11H13N | 159.23 g/mol

It is used in synthesis of cyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthesis of cyanine dyes.

Solubility
Soluble in chloroform, toluene or dichlorobenzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1640-39-7
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00005724
InChI Key FLHJIAFUWHPJRT-UHFFFAOYSA-N
Synonym 2,3,3-trimethylindolenine, 2,3,3-trimethyl-3 h-indole, 3 h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin
PubChem CID 15427
IUPAC Name 2,3,3-trimethyl-3H-indole
SMILES CC1=NC2=CC=CC=C2C1(C)C

Specifications

Melting Point 6°C to 8°C
Density 0.992
Boiling Point 228°C to 229°C
Flash Point 93°C
Refractive Index 1.547
Quantity 250 g
Beilstein 119740
Solubility Information Soluble in chloroform,toluene or dichlorobenzene.
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material 2,3,3-Trimethylindolenine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.