missing translation for 'onlineSavingsMsg'
Learn More

2',3',4,4',5-Pentachlorobiphenyl, TRC

Product Code. 30179937
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179937 100 mg 100mg
30180417 10 mg 10mg
30180397 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179937 Supplier Toronto Research Chemicals Supplier No. TRCP238195100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32598-14-4
Molecular Formula C12 H5 Cl5
Molecular Weight (g/mol) 326.43
Synonym 1,1'-Biphenyl, 2,3,3',4,4'-pentachloro-, Biphenyl, 2,3,3',4,4'-pentachloro- (8CI), 2,3,3',4,4'-Pentachloro-1,1'-biphenyl, 2,3,3',4,4'-Pentachlorobiphenyl, 2,3,4,3',4'-Pentachlorobiphenyl, 2',3',4,4',5-Pentachlorobiphenyl, 3,4,2',3',4'-Pentachlorobiphenyl, CB 105, PCB 105, Polychlorinated biphenyl-105, 2,3,3',4,4'-PentaCB
IUPAC Name 1,2,3-trichloro-4-(3,4-dichlorophenyl)benzene
SMILES Clc1ccc(cc1Cl)c2ccc(Cl)c(Cl)c2Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H5Cl5/c13-8-3-1-6(5-10(8)15)7-2-4-9(14)12(17)11(7)16/h1-5H
Formula Weight 323.8834
Chemical Name or Material PCB No. 105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.