missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzeneboronic acid, 97%

Product Code. 11479977
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479977 5 g 5g
11489977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479977 Supplier Thermo Scientific Alfa Aesar Supplier No. B22922.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1582-24-7 | C6H2BF5O2 | 211.88 g/mol

2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

Solubility
Miscible with alcohols, acetonitrile and dichloromethane.

Notes
Moisture Sensitive. Store in cool and dry conditions in well sealed containers. Keep container tightly sealed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1582-24-7
Molecular Formula C6H2BF5O2
Molecular Weight (g/mol) 211.88
MDL Number MFCD01074663
InChI Key VASOMTXTRMYSKD-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzeneboronic acid, pentafluorophenylboronic acid, perfluorophenylboronic acid, pentafluorobenzeneboronic acid, boronic acid, pentafluorophenyl, pentafluorophenyl boronic acid, 2,3,4,5,6-pentafluorophenyl boronic acid, boronic acid, b-2,3,4,5,6-pentafluorophenyl, pubchem22760, rarechem ah pb 0049
PubChem CID 2775922
IUPAC Name (2,3,4,5,6-pentafluorophenyl)boronic acid
SMILES OB(O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 250°C to 255°C
Quantity 5 g
Beilstein 3033960
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,acetonitrile and dichloromethane.
Formula Weight 211.88
Percent Purity 97%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.